Skip to Content
Merck
All Photos(1)

Key Documents

132489

Sigma-Aldrich

2-Chloropyrazine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$38.50
25 G
$60.10

$38.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$38.50
25 G
$60.10

About This Item

Empirical Formula (Hill Notation):
C4H3ClN2
CAS Number:
Molecular Weight:
114.53
Beilstein:
107019
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39162102
PubChem Substance ID:
NACRES:
NA.22

$38.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

153-154 °C (lit.)

density

1.283 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Clc1cnccn1

InChI

1S/C4H3ClN2/c5-4-3-6-1-2-7-4/h1-3H

InChI key

GELVZYOEQVJIRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

remainder water

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Wilke et al.
Journal of chromatography, 383(1), 77-83 (1986-11-28)
Diazohydroxide is a new antitumor agent being considered for clinical trial. A sensitive and specific assay for diazohydroxide in physiological media, plasma and blood has been developed based on conversion of diazohydroxide to 2-chloropyrazine in the presence of strong hydrochloric
Jeanne-Marie Bégouin et al.
The Journal of organic chemistry, 74(8), 3221-3224 (2009-03-25)
A cobalt-catalyzed cross-coupling of aryl halides with 2-chloropyrimidines or 2-chloropyrazines is reported in satisfactory to high yields. The key step of this procedure is the formation of aromatic organozinc reagents and their coupling with 2-chlorodiazines using the same cobalt halide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service