Skip to Content
Merck
All Photos(2)

Key Documents

131407

Sigma-Aldrich

1,2-Phenylenediacetic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2CO2H)2
CAS Number:
Molecular Weight:
194.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

150-152 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)Cc1ccccc1CC(O)=O

InChI

1S/C10H10O4/c11-9(12)5-7-3-1-2-4-8(7)6-10(13)14/h1-4H,5-6H2,(H,11,12)(H,13,14)

InChI key

MMEDJBFVJUFIDD-UHFFFAOYSA-N

General description

A coordination polymer with 1,2-Phenylenediacetic acid was synthesized and characterized by single crystal X-ray diffraction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, and luminescence of a novel lead (II) coordination polymer with 1, 2-phenylenediacetic acid.
Lei ZH, et al.
Inorganic Chemistry Communications, 13(12), 1383-1386 (2010)
Pengcheng Xing et al.
The Analyst, 144(8), 2656-2661 (2019-03-08)
Recently, the pollution and damage caused by antibiotics in water have aroused serious concerns. In this situation, it is extremely important to develop a highly effective approach to detect antibiotics in water. In this contribution, we built a Cd-MOF material

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service