Skip to Content
Merck
All Photos(1)

Key Documents

129186

Sigma-Aldrich

1,4-Hexadiene, mixture of cis and trans

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH2CH=CH2
CAS Number:
Molecular Weight:
82.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.411 (lit.)

bp

64-66 °C (lit.)

density

0.71 g/mL at 25 °C (lit.)

functional group

allyl

storage temp.

2-8°C

SMILES string

CC=CCC=C

InChI

1S/C6H10/c1-3-5-6-4-2/h3-4,6H,1,5H2,2H3

InChI key

PRBHEGAFLDMLAL-UHFFFAOYSA-N

Application

1,4-Hexadiene was used to prepare OH-adducts[1].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamad Al-Sheikhly et al.
Environmental science & technology, 40(9), 3082-3088 (2006-05-25)
Pulse radiolysis with spectrophotometric and conductometric detection was utilized to study the formation and reactions of radicals from benzene and dienes in aqueous solutions. The benzene OH adduct, *C6H6OH, reacts with O2 (k = 3 x 10(8) L mol(-1) s(-1))
Hexadiene.
American Industrial Hygiene Association journal, 43(1), B89-B90 (1982-01-01)
A versatile hexadiene synthesis by decarboxylative sp(3)-sp(3) coupling/cope rearrangement.
Shelli R Waetzig et al.
Angewandte Chemie (International ed. in English), 45(30), 4977-4980 (2006-07-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service