Skip to Content
Merck
All Photos(3)

Key Documents

128740

Sigma-Aldrich

2-Hydroxy-6-methylpyridine

97%

Synonym(s):

6-Methyl-2-pyridinol, 6-Methyl-2-pyridone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$74.00
25 G
$161.00

$74.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$74.00
25 G
$161.00

About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
Beilstein:
107073
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$74.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

157-159 °C (lit.)

SMILES string

Cc1cccc(O)n1

InChI

1S/C6H7NO/c1-5-3-2-4-6(8)7-5/h2-4H,1H3,(H,7,8)

InChI key

JEAVIRYCMBDJIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxy-6-methylpyridine reacts with carboxylic acids RCOOH to give an equilibrium mixture of products M(2)(O(2)CR)(n)(mhp)(4-n) where R = 2-thienyl and phenyl. mhp is the anion formed from deprotonation of 2-hydroxy-6-methylpyridine[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas J Brown et al.
Dalton transactions (Cambridge, England : 2003), (18)(18), 1793-1801 (2007-05-02)
The compounds M(2)(mhp)(4), where M = Mo or W and mhp is the anion formed from deprotonation of 2-hydroxy-6-methylpyridine, are shown to react with carboxylic acids RCOOH to give an equilibrium mixture of products M(2)(O(2)CR)(n)(mhp)(4-n) where R = 2-thienyl and
Feriel Boussema et al.
Biosensors & bioelectronics, 109, 20-26 (2018-03-11)
Two new inorganic-organic hybrid materials based on heteropolyoxometalates (POMs): (C4H10N)6[P2Mo18O62]·4H2O (P2Mo18) and (C6H8NO)4[H2P2W18O62]·6H2O (P2W18) are reported as mediators for electron transfer between FAD-dependent glucose dehydrogenase (FAD-GDH) and a multiwalled carbon nanotube (MWCNT) matrix for glucose biofuel cell and biosensor applications.
Emma Kirkman-Davis et al.
Dalton transactions (Cambridge, England : 2003), 49(39), 13693-13703 (2020-10-01)
A family of pyrazine-bridged, linear chain complexes of Cu(ii) of the formula [CuL2(H2O)2(pz)](ClO4)2 [pz = pyrazine; L = n-methyl-2(1H)-pyridone, n = 3 (1), 5 (2), and 6 (3)] has been prepared. Single-crystal X-ray diffraction shows six-coordinate, pyrazine-bridged chains with trans-pairs
Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service