Skip to Content
Merck
All Photos(1)

Key Documents

128414

Sigma-Aldrich

4′-Fluoropropiophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4C(O)C2H5
CAS Number:
Molecular Weight:
152.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.5059 (lit.)

density

1.096 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

SMILES string

CCC(=O)c1ccc(F)cc1

InChI

1S/C9H9FO/c1-2-9(11)7-3-5-8(10)6-4-7/h3-6H,2H2,1H3

InChI key

QIJNVLLXIIPXQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4′-Fluoropropiophenone was used in the preparation of 2-(4-fluorophenyl)-3-methylquinoxaline[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Padmavathy K, et al.
Tetrahedron Letters, 52(4) (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service