Skip to Content
Merck
All Photos(1)

Key Documents

127787

Sigma-Aldrich

Crotonoyl chloride

technical grade, 90%

Synonym(s):

(E)-Crotonoyl chloride, trans-Crotonoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCOCl
CAS Number:
Molecular Weight:
104.53
Beilstein:
506118
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical grade

Quality Level

Assay

90%

contains

1500 ppm hydroquinone as stabilizer

refractive index

n20/D 1.459 (lit.)

bp

120-123 °C (lit.)

density

1.091 g/mL at 25 °C (lit.)

functional group

acyl chloride

storage temp.

2-8°C

SMILES string

C\C=C\C(Cl)=O

InChI

1S/C4H5ClO/c1-2-3-4(5)6/h2-3H,1H3/b3-2+

InChI key

RJUIDDKTATZJFE-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Crotonoyl chloride was used in the synthesis of α,β-unsaturated ester[1].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Double asymmetric induction as a mechanistic probe: the doubly diastereoselective conjugate addition of enantiopure lithium amides to enantiopure α,β-unsaturated esters and enantiopure α,β-unsaturated hydroxamates.
Davies SG, et al.
Tetrahedron, 67(34), 6382-6403 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service