Skip to Content
Merck
All Photos(2)

Key Documents

127116

Sigma-Aldrich

N,N,N′-Triethylethylenediamine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$101.00

$101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$101.00

About This Item

Linear Formula:
(C2H5)2NCH2CH2NHC2H5
CAS Number:
Molecular Weight:
144.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$101.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.4311 (lit.)

bp

54-55 °C/13 mmHg (lit.)

density

0.804 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCNCCN(CC)CC

InChI

1S/C8H20N2/c1-4-9-7-8-10(5-2)6-3/h9H,4-8H2,1-3H3

InChI key

HDCAZTXEZQWTIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N,N′-triethylethylenediamine has been used to study the synthesis and characterization of N,N,N′-triethylethylenediamine by spectroscopic, magnetic, molar conductance and electrochemical measurements[1].

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Taha
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(6), 1373-1386 (2003-03-28)
Solvatochromic mixed ligand complexes of copper(II) with malonate and diamine derivatives, Cu(n)(RMal)(diam)(n)X(m) (where n=1 or 2, m=1-4, RMal, malonic acid (H(2)Mal), diethylmalonate (HDEtMal) or diethylethoxyethylenemalonate (DEtEMal), and diam, ethylenediamine (en), 1,3-propylenediamine (1,3-pn), N,N,N'-trimethylethylenediamine (Me(3)en), N,N,N'-triethylethylenediamine (Et(3)en), N,N,N',N'-tetramethylethylenediamine (Me(4)en), N,N,N',N'-tetramethylpropylenediamine (Me(4)pn)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service