Skip to Content
Merck
All Photos(2)

Key Documents

126268

Sigma-Aldrich

2,4-Dihydroxypyrimidine-5-carboxylic acid

95%

Synonym(s):

Isoorotic acid, Uracil-5-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$84.30

$84.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$84.30

About This Item

Empirical Formula (Hill Notation):
C5H4N2O4
CAS Number:
Molecular Weight:
156.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$84.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

mp

283 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)C1=CNC(=O)NC1=O

InChI

1S/C5H4N2O4/c8-3-2(4(9)10)1-6-5(11)7-3/h1H,(H,9,10)(H2,6,7,8,11)

InChI key

ZXYAAVBXHKCJJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dihydroxypyrimidine-5-carboxylic acid (Uracil-5-carboxylic acid) has been obtained from 5-formyluracil by the action of enzyme, thymine 7-hydroxylase[1]. It has been used to synthesize N1-alkylated uracil derivatives[2].

Application

2,4-Dihydroxypyrimidine-5-carboxylic acid (Uracil-5-carboxylic acid) has been used for the visual sensing of melamine (at parts-per-billion (ppb) level) by a highly sensitive analytical method based on Au nanoparticles[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

P M Shaffer et al.
Journal of bacteriology, 121(2), 648-655 (1975-02-01)
The experiments in this report involve the following series of reactions which were previously demonstrated with purified enzyme preparations from Neurospora crassa: thymidine a yields thymine ribonucleoside b yields thymine c yields 5-hydroxymethyluracil d yields 5-formyluracil e yields uracil-5-carboxylic acid
Raj Kumar Bera et al.
The Analyst, 136(8), 1644-1648 (2011-02-25)
A highly sensitive analytical method based on Au nanoparticles rationally tailored with recognition elements uracil-5-carboxylic acid (UCA) and 2,4,6-trinitrobenzenesulfonic acid (TNBS) for the visual sensing of melamine at the parts-per-billion (ppb) level is described. The tailored Au nanoparticles function as
Alessandro Accetta et al.
Journal of medicinal chemistry, 52(1), 87-94 (2008-12-17)
The synthesis of C5 linked uracil dimers was carried out according to a model developed in order to bind adenine in DNA. N1-Alkylated uracil derivatives were synthesized from isoorotic acid (uracil-5-carboxylic acid) or thymine. The carboxylic acid derivatives were condensed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service