Skip to Content
Merck
All Photos(1)

Key Documents

124079

5-Bromoisatin

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4BrNO2
CAS Number:
Molecular Weight:
226.03
Beilstein:
383760
MDL number:

Assay

95%

mp

247-252 °C (lit.)

SMILES string

Brc1ccc2NC(=O)C(=O)c2c1

InChI

1S/C8H4BrNO2/c9-4-1-2-6-5(3-4)7(11)8(12)10-6/h1-3H,(H,10,11,12)

InChI key

MBVCESWADCIXJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains isatin

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service