Skip to Content
Merck
All Photos(2)

Key Documents

122947

Sigma-Aldrich

4-Amino-9-fluorenone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO
CAS Number:
Molecular Weight:
195.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

138-140 °C (lit.)

functional group

ketone

SMILES string

Nc1cccc2C(=O)c3ccccc3-c12

InChI

1S/C13H9NO/c14-11-7-3-6-10-12(11)8-4-1-2-5-9(8)13(10)15/h1-7H,14H2

InChI key

FFWCEONGEXZNFU-UHFFFAOYSA-N

Application

4-Amino-9-fluorenone was used in a study on determination of major contaminants in water samples from gas plant by using particle beam interface LC-coupled to a single quadrupole mass spectrometer[1]. It was used as starting reagent for the synthesis of 4-methoxy-9-fluorenone[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Dei et al.
Bioorganic & medicinal chemistry, 9(10), 2673-2682 (2001-09-15)
Several ring-substituted derivatives of previously studied MDR inhibitors 2-(3,4-dimethoxyphenyl)-5-(9-fluorenylamino)-2-(methylethyl)pentanenitrile and 2-(3,4-dimethoxyphenyl)-5-[(9-fluorenyl)-N-methylamino]-2-(methylethyl)pentanenitrile have been synthesised and studied with the aim of optimising activity and selectivity. The results show that MDR inhibition is scarcely sensitive to modulation of the electronic properties of
Particle beam LC/MS screening of polar compounds of contaminated ground water samples from a former gas plant.
Edler BC, et al.
Fresenius Journal of Analytical Chemistry, 359(3), 288-292 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service