Skip to Content
Merck
All Photos(1)

Key Documents

120189

Sigma-Aldrich

3,4-Dichlorophenoxyacetic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OCH2CO2H
CAS Number:
Molecular Weight:
221.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

138-140 °C (lit.)

SMILES string

OC(=O)COc1ccc(Cl)c(Cl)c1

InChI

1S/C8H6Cl2O3/c9-6-2-1-5(3-7(6)10)13-4-8(11)12/h1-3H,4H2,(H,11,12)

InChI key

SNYRXHULAWEECU-UHFFFAOYSA-N

General description

3,4-Dichlorophenoxyacetic acid is a phenoxyacetic herbicide.

Application

3,4-Dichlorophenoxyacetic acid was used in the synthesis of layered organic-inorganic nanohybrid material, zinc-aluminum-3,4-dicholorophenoxyacetate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ludovic Lardet et al.
Tree physiology, 29(2), 291-298 (2009-02-11)
We compared embryogenic capacities of integument explants excised from three sources of the Hevea brasiliensis (Müll. Arg.) mature genotype PB 260. The three sources were 17-year-old (BT 86) and 7-year-old (BT 96) budded trees and 7-year-old emblings (EM 96). The
Sheikh Ahmad Izaddin Sheikh Mohd Ghazali et al.
Nanoscale research letters, 8(1), 362-362 (2013-08-24)
A new layered organic-inorganic nanohybrid material, zinc-aluminum-3,4-dicholorophenoxyacetate (N3,4-D) in which an agrochemical, 3,4-dichlorophenoxyacetic acid (3,4-D), is intercalated into zinc-aluminum-layered double hydroxide (ZAL), was synthesized by coprecipitation method. A well-ordered nanomaterial was formed with a percentage loading of 53.5% (w/w). Due
D E Lynch et al.
Acta crystallographica. Section C, Crystal structure communications, 56(Pt 12), 1478-1479 (2000-12-19)
The 1:1 organic salt of the title compound, C(7)H(6)ClN(2)O(+). C(8)H(5)Cl(2)O(3)(-) or [(2-ABOX)(3,4-D)], comprises the two constituent molecules associated by an R(2)(2)(8) graph-set interaction through the carboxylate group of 3,4-D across the protonated N/N sites of 2-ABOX [N.O 2.546 (3) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service