Skip to Content
Merck
All Photos(1)

Key Documents

117609

Sigma-Aldrich

Cyclopentadienylmolybdenum(II) tricarbonyl, dimer

98%

Synonym(s):

Cyclopentadienyltricarbonylmolybdenum dimer, Hexacarbonylbis(η5 -cyclopentadienyl)dimolybdenum(I), Hexacarbonylbis(cyclopentadienyl)dimolybdenum

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$79.40

$79.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$79.40

About This Item

Empirical Formula (Hill Notation):
C16H10Mo2O6
CAS Number:
Molecular Weight:
490.13
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$79.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

core: molybdenum
reagent type: catalyst

mp

222 °C (dec.) (lit.)

SMILES string

[Mo][Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.6CO.2Mo/c2*1-2-4-5-3-1;6*1-2;;/h2*1-5H;;;;;;;;

InChI key

XCUPBPUEWLTPMQ-UHFFFAOYSA-N

Application

  • Group 6 Carbonyl Complexes of N, O, P-ligands: Reviews the use of cyclopentadienyl molybdenum(II) dicarbonyl and tricarbonyl complexes as precursors for catalysts in olefin epoxidation. (SM Bruno, AA Valente, IS Gonçalves, 2023).
  • Monitoring Equilibrium Reaction Dynamics of a Nearly Barrierless Molecular Rotor: Describes using cyclopentadienylmolybdenum tricarbonyl dimer in ultrafast vibrational echo experiments to observe dynamic processes in barrierless molecular rotors. (IA Nilsen, DG Osborne, AM White, JM Anna, 2014).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service