Skip to Content
Merck
All Photos(1)

Key Documents

116173

Sigma-Aldrich

2-Bromo-3-pyridinol

99%

Synonym(s):

2-Bromo-3-hydroxypyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4BrNO
CAS Number:
Molecular Weight:
174.00
Beilstein:
109829
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

185-188 °C (lit.)

SMILES string

Oc1cccnc1Br

InChI

1S/C5H4BrNO/c6-5-4(8)2-1-3-7-5/h1-3,8H

InChI key

YKHQFTANTNMYPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromo-3-pyridinol has been used in the preparation of 2-bromo-4,6-diiodo-3-pyridinol. It has also been used in the total synthesis of pterocellin A by reacting with kojic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Halogenation of Pyridinols using Bis(sym-collidine)iodine(I) and Bis(sym-collidine)bromine(I) hexafluorophosphate.
Tetrahedron Letters, 38(14), 2467-2470 (1997)
Meaghan M O'Malley et al.
Organic letters, 8(12), 2651-2652 (2006-06-02)
The first total synthesis of pterocellin A (1) was achieved in 10 linear steps from commercially available kojic acid (6) and 2-bromo-3-pyridinol (11) in a convergent sequence. The key constructive steps are a directed lithiation to couple two pyridines and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service