Skip to Content
Merck
All Photos(1)

Key Documents

115878

Sigma-Aldrich

2,3-Dimethoxytoluene

99%

Synonym(s):

3-Methylveratrole

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$226.00

$226.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$226.00

About This Item

Linear Formula:
CH3C6H3(OCH3)2
CAS Number:
Molecular Weight:
152.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$226.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

refractive index

n20/D 1.514 (lit.)

bp

202-203 °C (lit.)

density

1.025 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(C)c1OC

InChI

1S/C9H12O2/c1-7-5-4-6-8(10-2)9(7)11-3/h4-6H,1-3H3

InChI key

WMXFNCKPYCAIQW-UHFFFAOYSA-N

Related Categories

Application

2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darío A Bianchi et al.
Journal of agricultural and food chemistry, 52(7), 1923-1927 (2004-04-01)
The synthesis of 6,7-dimethoxy-4-hydroxyisochroman-3-one 10 from 2,3-dimethoxytoluene (11) via glyoxylate 12 is reported. Compound 10 strongly inhibited vegetative growth of tobacco plants, whereas developmental patterns (protein levels, protein profile, pigments, and chlorophylls) were not affected. Morphological changes were observed in
Olga V Zalomaeva et al.
Dalton transactions (Cambridge, England : 2003), 46(16), 5202-5209 (2017-04-01)
The selective oxidation of methoxy/methyl-substituted arenes to the corresponding benzoquinones has been first realized using aqueous hydrogen peroxide as a green oxidant, acid tetrabutylammonium salts of the γ-Keggin divanadium-substituted phosphotungstate [γ-PW

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service