Skip to Content
Merck
All Photos(1)

Key Documents

113832

Sigma-Aldrich

3,5-Diaminobenzoic acid dihydrochloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2N)2C6H3CO2H·2HCl
CAS Number:
Molecular Weight:
225.07
Beilstein:
3703362
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

reaction suitability

reaction type: solution phase peptide synthesis

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl[H].Cl[H].Nc1cc(N)cc(c1)C(O)=O

InChI

1S/C7H8N2O2.2ClH/c8-5-1-4(7(10)11)2-6(9)3-5;;/h1-3H,8-9H2,(H,10,11);2*1H

InChI key

IGPLRBRBTUNCRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Uses include the fluorometric microdetermination of DNA.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interference of Bis-Tris buffer with the diaminobenzoic acid fluorescence assay used to quantify DNA.
W E Schy et al.
Mutation research, 226(4), 263-266 (1989-08-01)
J Beckmann et al.
Acta diabetologica latina, 18(1), 51-57 (1981-01-01)
DNA content seems to be an ideal reference parameter for data on secretory function or metabolism of pancreatic islets. The approved fluorometric DNA assay with diaminobenzoic acid (DABA) of Kissane and Robins comprises repeated ethanol extractions of the tissue for
Céline Douat-Casassus et al.
Journal of the American Chemical Society, 126(25), 7817-7826 (2004-06-24)
Peptide dendrimers incorporating 3,5-diaminobenzoic acid 1 as a branching unit (B) were prepared by solid-phase synthesis of ((Ac-A(3))(2)B-A(2))(2)B-Cys-A(1)-NH(2) followed by disulfide bridge formation. Twenty-one homo- and heterodimeric dendrimers were obtained by permutations of aspartate, histidine, and serine at positions A(1)
[Improved fluorometric method for DNA microanalysis].
M M Vilenchik et al.
Izvestiia Akademii nauk SSSR. Seriia biologicheskaia, (2)(2), 217-222 (1984-03-01)
R J Wilkins et al.
Analytical biochemistry, 136(2), 301-308 (1984-02-01)
Procedures for collecting DNA from dilute solutions as small spots of the insoluble cetyltrimethylammonium salt deposited on either glass fiber or Millipore filters are described. This method of concentration facilitates the fluorescent assay of DNA, either by a microreaction in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service