Skip to Content
Merck
All Photos(3)

Key Documents

112747

Sigma-Aldrich

N-Acetylsulfanilyl chloride

98%

Synonym(s):

4-Acetamidobenzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$86.60
1 KG
$209.00

$86.60


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$86.60
1 KG
$209.00

About This Item

Linear Formula:
4-(CH3CONH)C6H4SO2Cl
CAS Number:
Molecular Weight:
233.67
Beilstein:
746676
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$86.60


Available to ship on17 April 2025Details


Request a Bulk Order

Assay

98%

mp

142-145 °C (dec.) (lit.)

functional group

amide

SMILES string

CC(=O)Nc1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C8H8ClNO3S/c1-6(11)10-7-2-4-8(5-3-7)14(9,12)13/h2-5H,1H3,(H,10,11)

InChI key

GRDXCFKBQWDAJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-hydroxyproline has been derivatized with N-acetylsulfanilyl chloride and 5-chlorovaleric acid during the synthesis of the haptens HP1 and HP2[1].

Application

N-Acetylsulfanilyl chloride is used in the synthesis of N1,N1-dimethylsulfanilamide as the internal standard in the reaction. The reaction is used to determine tramadol and its O-desmethylated metabolite in blood plasma[2].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li L Wang et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(1), 9-15 (2012-10-04)
China's government has prohibited the addition of simply hydrolyzed animal protein from solid leather waste into milk. The objective of this study was to produce a monoclonal antibody against L-hydroxyproline, a special amino acid in hydrolyzed animal protein. L-hydroxyproline was
Ning Li et al.
European journal of medicinal chemistry, 155, 531-544 (2018-06-18)
Ten novel symmetric 3,5-bis(arylidene)-4-piperidone derivatives (BAPs, 1-10) and fourteen dissymmetric BAPs (11-24) were synthesized and evaluated the cytotoxicity. All of the compounds have been screened for their anti-inflammatory activity characterized by evaluating their inhibitory effects on LPS-induced IL-6, TNF-α secretion.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service