Skip to Content
Merck
All Photos(1)

Key Documents

10919

Sigma-Aldrich

Fmoc isothiocyanate

≥98.0% (CHN)

Synonym(s):

9-Fluorenylmethoxycarbonyl isothiocyanate, 9-Fluorenylmethyl isothiocyanatoformate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$213.00
5 G
$775.00

$213.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$213.00
5 G
$775.00

About This Item

Empirical Formula (Hill Notation):
C16H11NO2S
CAS Number:
Molecular Weight:
281.33
Beilstein:
7930648
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$213.00


Check Cart for Availability

Request a Bulk Order

Assay

≥85% (coupling to amines)
≥98.0% (CHN)

solubility

ethanol: soluble

fluorescence

λex 264 nm; λem 313 nm

application(s)

peptide synthesis

functional group

Fmoc
amine
isothiocyanate

storage temp.

−20°C

SMILES string

O=C(OCC1c2ccccc2-c3ccccc13)N=C=S

InChI

1S/C16H11NO2S/c18-16(17-10-20)19-9-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15H,9H2

InChI key

DHMYULZVFHHEHE-UHFFFAOYSA-N

Application

Fmoc isothiocyanate can be used:
  • As a starting material in the preparation of biologically relevant pharmacophores named N-aryl-N′-carboalkoxy guanidines.[1]
  • In one of the intermediate steps for the synthesis of N-aryl-N-thiazolyl derivatives.[2]
  • To synthesize cyclic isothiourea derivatives as potent neuropeptide Y (NPY) Y1 receptor antagonists.[3]
  • To prepare 2-aminothiazoles,[4] aminobenz-imidazole conjugated thiazoles,[5] and thiazole derived cyclopeptides[6].

Other Notes

Reagent for the synthesis of substituted thioureas from amines[7]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diversity-oriented synthesis of N-aryl-N-thiazolyl compounds
Nefzi A and Arutyunyan S
Tetrahedron Letters, 51(37), 4797-4800 (2010)
Solid-Phase Synthesis of 2-Aminothiazoles.
Patrick C. Kearney et al.
The Journal of organic chemistry, 63(1), 196-200 (2001-10-25)
Solid-Phase Synthesis of N-Aryl-N′-Carboalkoxy Guanidines by the Mitsunobu Reaction of Fmoc-Guanidines
Robinson DE, et al.
Synthetic Communications, 34(15), 2743-2749 (2004)
Two-step hantzsch based macrocyclization approach for the synthesis of thiazole-containing cyclopeptides
Nefzi A, et al.
The Journal of Organic Chemistry, 75(22), 7939-7941 (2010)
Solid-phase synthesis of 2-aminothiazoles
Kearney PC, et al.
The Journal of Organic Chemistry, 63(1), 196-200 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service