Skip to Content
Merck
All Photos(1)

Key Documents

10899

Sigma-Aldrich

Methyl 3-oxoheptanoate

produced by Wacker Chemie AG, Burghausen, Germany, ≥96.0% (GC)

Synonym(s):

VEM, Methyl valerylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COCH2COOCH3
CAS Number:
Molecular Weight:
158.19
Beilstein:
1761380
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

produced by Wacker Chemie AG, Burghausen, Germany

Assay

≥96.0% (GC)

manufacturer/tradename

Wacker Chemie AG

bp

90-91 °C/15 hPa (lit.)

density

0.994 g/mL at 20 °C (lit.)

SMILES string

CCCCC(=O)CC(=O)OC

InChI

1S/C8H14O3/c1-3-4-5-7(9)6-8(10)11-2/h3-6H2,1-2H3

InChI key

CZTKGERSDUGZPQ-UHFFFAOYSA-N

General description

Methyl 3-oxoheptanoate undergoes Pechmann condensation with 2-methyl resorcinol using H2SO4 condensing agent to form 7-hydroxy-8-methyl coumarins.

Application

Methyl 3-oxoheptanoate was used in the synthesis of 3(R,S)-hydroxyheptanoic acid.

Other Notes

prices for bulk quantities on request

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F

Flash Point(C)

95 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria E McCoy et al.
Scientific reports, 9(1), 6420-6420 (2019-04-25)
Ancient protein analysis is a rapidly developing field of research. Proteins ranging in age from the Quaternary to Jurassic are being used to answer questions about phylogeny, evolution, and extinction. However, these analyses are sometimes contentious, and focus primarily on
Abdullah Sisik et al.
Obesity surgery, 29(2), 444-450 (2018-09-29)
The effect of local anesthetic applications to trocar sites on postoperative pain control has been studied many times, and different results have been obtained. We planned a controlled study evaluating the effect of bupivacaine administration and other contributing factors on
Petar Djuric et al.
Toxins, 12(6) (2020-05-31)
Increased oxidative stress is a hallmark of end-stage renal disease. Hemodialysis (HD) patients lacking glutathione transferase M1 (GSTM1) enzyme activity exhibit enhanced oxidative DNA damage and higher mortality rate than those with active GSTM1 enzyme. To our knowledge, this is
G R Markby et al.
BMC veterinary research, 16(1), 350-350 (2020-09-25)
Almost all elderly dogs develop myxomatous mitral valve disease by the end of their life, but the cavalier King Charles spaniel (CKCS) has a heightened susceptibility, frequently resulting in death at a young age and suggesting that there is a
Mastour A Alhamami et al.
Optometry and vision science : official publication of the American Academy of Optometry, 94(2), 137-149 (2016-11-16)
To investigate whether cysts in diabetic macular edema are better visualized in the red channel of color fundus camera images, as compared with the green channel, because color fundus camera screening methods that emphasize short-wavelength light may miss cysts in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service