Skip to Content
Merck
All Photos(1)

Key Documents

106550

Sigma-Aldrich

meso-1,2-Dibromo-1,2-diphenylethane

≥97%

Synonym(s):

Stilbene dibromide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$160.00
100 G
$497.00

$160.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$160.00
100 G
$497.00

About This Item

Linear Formula:
C6H5CH(Br)CH(Br)C6H5
CAS Number:
Molecular Weight:
340.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$160.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97%

form

solid

mp

241 °C (dec.) (lit.)

functional group

bromo
phenyl

SMILES string

Br[C@H]([C@H](Br)c1ccccc1)c2ccccc2

InChI

1S/C14H12Br2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-14H/t13-,14+

InChI key

GKESIQQTGWVOLH-OKILXGFUSA-N

General description

meso-1,2-Dibromo-1,2-diphenylethane undergoes electrochemical dehalogenation in acetonitrile[1]. It is reduced by electrogenerated C60−3[2].

Application

meso-1,2-Dibromo-1,2-diphenylethane(meso-SBr2) was used to study the reaction of ± and meso-SBr2 with 9-substituted fluorenide ions in dimethyl sulfoxide[3].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debromination of meso-and (?)-1, 2-Dibromo-1, 2-diphenylethane by 9-Substituted Fluorenide Ions: Correlation between Stereochemical Results and Redox Potentials.
Lund T, et al.
Acta Chemica Scandinavica, 47, 877-877 (1993)
Electro-organic chemistry of fullerenes. Part 1. Indirect cathodic reduction of vic-dihalides and perfluoroalkyl halides using C60 as mediator. Cyclic voltammetric study and preparative-scale electrolysis.
Fuchigami T, et al.
Journal of Electroanalytical Chemistry, 411(1), 115-119 (1996)
Reduction of Vicinal Dihalides. I. The Electrochemical Reduction of meso and (?)-1, 2-Dibromo-1, 2-diphenylethane.
Fawell P, et al.
Australian Journal of Chemistry, 43(8), 1421-1430 (1990)

Questions

1–6 of 6 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Where can I find product similar to meso-1,2-Dibromo-1,2-diphenylethane offered by Sigma-Aldrich?

    1 answer
    1. The Sigma-Aldrich website provides many resources for locating products with similar structural characteristics. At the bottom of the product information page for product 106550, there are links to "Related Categories". These links including listings of Sigma-Aldrich products that fall in the same category as meso-1,2-Dibromo-1,2-diphenylethane. Alternatively, Sigma-Aldrich offers an online catalog that provides organized listings of products useful for chemical synthesis, grouped by similar characteristics.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. Is Stilbene dibromide, Product 106550, optically active?

    1 answer
    1. Generally, chiral molecules are optically active, rotating polarized light. Meso compounds, such as 106550, are unique in that there are chiral carbons in the molecule yet they are not optically active. meso-1,2-Dibromo-1,2-diphenylethane has chiral carbons that are virtually identical but the arrangement of the groups on this carbon are different; the result of these opposing effects is a net optical rotation of zero.

      Helpful?

  6. In what solvents is Stilbene dibromide, Product 106550, soluble?

    1 answer
    1. Sigma-Aldrich does not currently test this product for solubility in a particular solvent. However, our quality control does solubilize this product with acetonitrile (with sonication) for analysis by HPLC at a concentration of 0.5mg/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service