Skip to Content
Merck
All Photos(1)

Documents

102164

Sigma-Aldrich

3,4-Dihydroxy-DL-phenylalanine

97%

Synonym(s):

3-(3,4-Dihydroxyphenyl)-DL-alanine, DL-3-Hydroxytyrosine, DL-DOPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
197.19
Beilstein:
3204800
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

mp

270-272 °C (lit.)
290 °C (dec.) (lit.)

SMILES string

NC(Cc1ccc(O)c(O)c1)C(O)=O

InChI

1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)

InChI key

WTDRDQBEARUVNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven L Saville et al.
Nanoscale, 5(5), 2152-2163 (2013-02-08)
It has been recently reported that for some suspensions of magnetic nanoparticles the transverse proton relaxation rate, R(2), is dependent on the time that the sample is exposed to an applied magnetic field. This time dependence has been linked to
Luis García-Fernández et al.
Advanced materials (Deerfield Beach, Fla.), 25(4), 529-533 (2012-11-10)
Inspired by the amino acid 2-chloro-4,5-dihydroxyphenylalanine (Cl-DOPA), present in the composition of the proteinaceous glue of the sandcastle worm Phragmatopoma californica, a simple strategy is presented to confer antifouling properties to polymer surfaces using (but not releasing) a bioinspired biocide.
Meng-Yao Lu et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(1), 42-49 (2012-12-06)
Neuroblastic tumors are childhood neoplasms that possess amino acid decarboxylase (AADC) activity and can theoretically be imaged by (18)F-fluorodihydroxyphenylalanine ((18)F-FDOPA) PET, a new diagnostic tool for neuroendocrine tumors. In this study, we explored the accuracy and clinical role of (18)F-FDOPA
Dongyeop X Oh et al.
Biotechnology progress, 29(2), 505-512 (2013-01-16)
Chitosan is one of the most widely used structural polymers for biomedical applications because it has many favorable properties. However, one of the most critical drawbacks regarding the use of chitosan as a biomedical material is its poor mechanical properties
Karlo J Lizarraga et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(1), 30-36 (2013-10-30)
Brain metastases are frequently treated with radiation. It is critical to distinguish recurrent or progressive brain metastases (RPBM) from late or delayed radiation injury (LDRI). The purpose of this study was to examine the diagnostic accuracy as well as the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service