Skip to Content
Merck
All Photos(1)

Key Documents

AABH97CE8B1E

AA BLOCKS, INC.

ethyl [(pentan-3-yl)carbamoyl]formate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
187.24
MDL number:

Iupac Name

ethyl [(1-ethylpropyl)amino](oxo)acetate

Inchi Code

1S/C9H17NO3/c1-4-7(5-2)10-8(11)9(12)13-6-3/h7H,4-6H2,1-3H3,(H,10,11)

InChI key

GSABFEQLGUXMPD-UHFFFAOYSA-N

storage temp.

2~8 C

Purity

95%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/349442-60-0

Synonym

Ethyl[(1-ethylpropyl)amino](oxo)acetate

MSDS

https://www.aablocks.com/downloadSds?cas=349442-60-0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library