Skip to Content
Merck
All Photos(1)

Key Documents

S9009

Sigma-Aldrich

Sar-Pro-Arg p-nitroanilide dihydrochloride

thrombin substrate, ≥97% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30N8O5 · 2HCl
CAS Number:
Molecular Weight:
535.42
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Sar-Pro-Arg p-nitroanilide dihydrochloride, thrombin substrate

Quality Level

Assay

≥97% (HPLC)

form

powder

solubility

water: 25 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

Cl.CNCC(=O)N1CCCC1C(=O)NC(CCCNC(N)=N)C(=O)Nc2ccc(cc2)N(=O)=O

InChI

1S/C20H30N8O5.ClH/c1-23-12-17(29)27-11-3-5-16(27)19(31)26-15(4-2-10-24-20(21)22)18(30)25-13-6-8-14(9-7-13)28(32)33;/h6-9,15-16,23H,2-5,10-12H2,1H3,(H,25,30)(H,26,31)(H4,21,22,24);1H

InChI key

KMSNYOSWLIQVAN-UHFFFAOYSA-N

Substrates

Chromogenic substrate for thrombin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Duncan et al.
Clinical chemistry, 31(6), 853-855 (1985-06-01)
Automated procedures involving a chromogenic substrate sensitive to thrombin-sarcosine-Pro-Arg p-nitroanilide were compared with conventional tests for prothrombin times and activated partial thromboplastin times (APTT) and with specific assays for factors V, VII, VIII, IX, X, XI, and XII. The reproducibility
Usman Bacha et al.
Chemical biology & drug design, 72(1), 34-49 (2008-07-10)
Coronaviruses comprise a large group of RNA viruses with diverse host specificity. The emergence of highly pathogenic strains like the SARS coronavirus (SARS-CoV), and the discovery of two new coronaviruses, NL-63 and HKU1, corroborates the high rate of mutation and

Protocols

Thrombin is an endolytic serine protease that selectively cleaves the Arg–Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service