Skip to Content
Merck
All Photos(2)

Key Documents

797294

Sigma-Aldrich

1,1,2,3,4,5-Hexaphenyl-1H-silole

Synonym(s):

1,1,2,3,4,5-Hexaphenylsilacyclopenta-2,4-diene, 1,1,2,3,4,5-Hexaphenylsilacyclopentadiene, 1,1,2,3,4,5-Hexaphenylsilole, HPS, Hexaphenylsilacyclopenta-2,4-diene, Hexaphenylsilacyclopentadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H30Si
CAS Number:
Molecular Weight:
538.75
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

188-193 °C

SMILES string

C1(C2=CC=CC=C2)=C(C3=CC=CC=C3)C(C4=CC=CC=C4)=C(C5=CC=CC=C5)[Si]1(C6=CC=CC=C6)C7=CC=CC=C7

InChI

1S/C40H30Si/c1-7-19-31(20-8-1)37-38(32-21-9-2-10-22-32)40(34-25-13-4-14-26-34)41(35-27-15-5-16-28-35,36-29-17-6-18-30-36)39(37)33-23-11-3-12-24-33/h1-30H

InChI key

QAKMXYFDVPDIPT-UHFFFAOYSA-N

General description

HPS is an aggregation-induced emission (AIE) material for use in the OLED emitting layer. Photolysis and thermolysis of 1,4,5,6,7,7-hexaphenyl-7-silabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride (XIb) may lead to the formation of hexaphenylsilacyclopentadiene.

Application

Hexaphenylsilacyclopentadiene may undergo π4s+π2s cycloaddition with ethyl acrylate to form bicyclosilaheptene. The same study reports similar cycloaddition reactions with various dienophiles. The reaction of hexaphenylsilacyclopentadiene with perbenzoic acid yields a mixture of tetraphenylfuran and cis-dibenzoylstilbene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Symmetry allowed ?4s+?2s additions silacyclopentadienes
Balasubramanian R and George MV
Tetrahedron null
Peracid oxidation and photooxygenation reactions of silacyclopentadienes,
Mahajan MP, et al.
Tetrahedron, 1549-1553 null
Photochemical and thermal transformations of 7-silabicyclo[2.2.1]Heptadiene and 7-silabicyclo[2.2.1]heptene systems
Balasubramanian R and George MV
Journal of Organometallic Chemistry, 131-139 null
Peracid oxidation and photooxygenation reactions of silacyclopentadienes,
Mahajan MP, et al.
Tetrahedron null
Symmetry allowed ?4s+?2s additions silacyclopentadienes
Balasubramanian R and George MV
Tetrahedron, 2395-2404 null

Articles

AIE Luminogens - Family of new materials with multifaceted functionalities. Find some representative applications of the AIEgens

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service