Merck
  • Ambient level volatile organic compound (VOC) monitoring using solid adsorbents--recent US EPA studies.
Ambient level volatile organic compound (VOC) monitoring using solid adsorbents--recent US EPA studies.
Journal of environmental monitoring : JEM (2002-10-29)
William A McClenny, Karen D Oliver, Henry H Jacumin, E Hunter Daughtrey
ABSTRACT
Ambient air spiked with 1-10 ppbv concentrations of 41 toxic volatile organic compounds (VOCs) listed in US Environmental Protection Agency (EPA) Compendium Method TO-14A was monitored using solid sorbents for sample collection and a Varian Saturn 2000 ion trap mass spectrometer for analysis. The adsorbent was a combination of graphitic carbon and a Carboxen-type carbon molecular sieve. The method detection limits (MDLs) for 11 samples were typically 0.5 parts per billion by volume (ppbv) and lower except for bromomethane and chloromethane, both of which exhibited breakthrough. Thirty-day sample storage on the sorbents resulted in less than a 20% change for most compounds, and water management was required for humid samples to avoid major anomalous decreases in response during analyses. The adsorbent-based system, a system using canister-based monitoring, and a semi-continuous automated GC/MS (autoGC) monitoring system with a Tenax GR/Carbotrap B/Carbosieve S-III adsorbent preconcentrator were compared using spiked ozone concentrations as a variable. In this comparison, the target compounds included a number of n-aldehydes as well as those listed in TO-14A. The effects of ozone on the TO-14A compounds were relatively minor with the exception of negative artifacts noted for styrene and 1,1,2,2-tetrachloroethane. However, a small, systematic decrease in response was evident for a number of aromatic VOCs and 1,1,2,2-tetrachloroethane when ozone was increased from 50 to 300 ppbv. Method averages for multiple runs under the same conditions were typically within +0.25 ppbv of their mean for most compounds. For n-aldehydes, strong positive artifacts using the autoGC preconcentrator and strong negative artifacts for the canister-based and carbon sorbent approaches caused major disagreement among methods. These artifacts were mostly eliminated by using MnO2 ozone scrubbers, although loss of the n-aldehydes for all methods occurred after a single sample collection of 1 h duration, apparently due to the interaction of the n-aldehydes and products of the O3, MnO2 reaction on the scrubber.
Product Description
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
Carbotrap® 349, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4 in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® X, 20-40 mesh, bottle of 10 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber