Merck
  • Determination of ethane, pentane and isoprene in exhaled air using a multi-bed adsorbent and end-cut gas-solid chromatography.
Determination of ethane, pentane and isoprene in exhaled air using a multi-bed adsorbent and end-cut gas-solid chromatography.
The Analyst (2002-12-12)
Mona Lärstad, Carina Loh, Göran Ljungkvist, Anna-Carin Olin, Kjell Torén
ABSTRACT
A method for the determination of exhaled ethane, pentane and isoprene was developed and validated. The method was based on pre-concentration of the analytes on a multi-bed solid adsorbent tube containing Tenax TA, Carboxen 569 and Carboxen 1000, thermal desorption and gas chromatography (GC) with flame ionisation detection (FID). A pre-column in an end-cut GC system was used to avoid problems with water and strongly retained substances. The detection limits were 5, 2 and 6 pmol per sample for ethane, pentane and isoprene, respectively, using a sample volume of 500 ml. The linearity was good for all analytes with correlation coefficients exceeding 0.999. The repeatability for exhaled air samples was 7, 10 and 12% for ethane, pentane and isoprene, respectively. Analysis of a certified reference material of ethane and pentane did not differ significantly from the certified values. Ethane and pentane levels were stable up to six days of storage in sample tubes. Isoprene levels were not stable during storage in the sample tubes used here, but using Carbopack X instead of Carboxen 569, levels were stable up to two days. The levels of exhaled ethane, pentane and isoprene in healthy subjects (n = 4) were 8.1+/-5.8 pmol l(-1), 11+/-5.8 pmol l(-1) and 2.4+/-0.90 mnol l(-1), respectively. The method could, with minor modifications, be used to determine other low-molecular hydrocarbons in exhaled air as well.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carbopack Adsorbent, matrix Carbopack X, 120-400 mesh, bottle of 50 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler