Merck
  • Survival motor neuron gene 2 silencing by DNA methylation correlates with spinal muscular atrophy disease severity and can be bypassed by histone deacetylase inhibition.
Survival motor neuron gene 2 silencing by DNA methylation correlates with spinal muscular atrophy disease severity and can be bypassed by histone deacetylase inhibition.
Human molecular genetics (2008-10-31)
Jan Hauke, Markus Riessland, Sebastian Lunke, Ilker Y Eyüpoglu, Ingmar Blümcke, Assam El-Osta, Brunhilde Wirth, Eric Hahnen
ABSTRACT
Spinal muscular atrophy (SMA), a common neuromuscular disorder, is caused by homozygous absence of the survival motor neuron gene 1 (SMN1), while the disease severity is mainly influenced by the number of SMN2 gene copies. This correlation is not absolute, suggesting the existence of yet unknown factors modulating disease progression. We demonstrate that the SMN2 gene is subject to gene silencing by DNA methylation. SMN2 contains four CpG islands which present highly conserved methylation patterns and little interindividual variations in SMN1-deleted SMA patients. The comprehensive analysis of SMN2 methylation in patients suffering from severe versus mild SMA carrying identical SMN2 copy numbers revealed a correlation of CpG methylation at the positions -290 and -296 with the disease severity and the activity of the first transcriptional start site of SMN2 at position -296. These results provide first evidence that SMN2 alleles are functionally not equivalent due to differences in DNA methylation. We demonstrate that the methyl-CpG-binding protein 2, a transcriptional repressor, binds to the critical SMN2 promoter region in a methylation-dependent manner. However, inhibition of SMN2 gene silencing conferred by DNA methylation might represent a promising strategy for pharmacologic SMA therapy. We identified histone deacetylase (HDAC) inhibitors including vorinostat and romidepsin which are able to bypass SMN2 gene silencing by DNA methylation, while others such as valproic acid and phenylbutyrate do not, due to HDAC isoenzyme specificities. These findings indicate that DNA methylation is functionally important regarding SMA disease progression and pharmacological SMN2 gene activation which might have implications for future SMA therapy regimens.
Sigma-Aldrich
Valproic acid sodium salt, 98%
Sigma-Aldrich
Zebularine, A cell-permeable chemically stable cytidine analog that displays antitumor properties.
Sigma-Aldrich
Sodium 4-Phenylbutyrate, A novel anti-neoplastic agent and transcriptional regulator.
Sigma-Aldrich
Histone Deacetylase Inhibitor III, The Histone Deacetylase Inhibitor III, also referenced under CAS 251456-60-7, controls the biological activity of Histone Deacetylase. This small molecule/inhibitor is primarily used for Cell Structure applications.
Sigma-Aldrich
Scriptaid, ≥95%, solid
Sigma-Aldrich
Oxamflatin, ≥98% (HPLC), solid