Skip to Content
Merck
All Photos(1)

Key Documents

567616

Sigma-Aldrich

Sodium 4-Phenylbutyrate

A novel anti-neoplastic agent and transcriptional regulator.

Synonym(s):

Sodium 4-Phenylbutyrate, SPB, PBNa, 4PBA, 4-Phenylbutyrate, Na, BCKD Kinase Inhibitor I, Branched-Chain α-Ketoacid Dehydrogenase Kinase Inhibitor I, BDK Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€98.30

€98.30


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€98.30

About This Item

Empirical Formula (Hill Notation):
C10H11NaO2
CAS Number:
Molecular Weight:
186.18
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.77

€98.30


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to slightly yellow

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[O-]C(=O)CCCc1ccccc1

InChI

1S/C10H12O2.Na/c11-10(12)8-4-7-9-5-2-1-3-6-9;/h1-3,5-6H,4,7-8H2,(H,11,12);/q;+1/p-1

InChI key

VPZRWNZGLKXFOE-UHFFFAOYSA-M

General description

A novel anti-neoplastic agent and transcriptional regulator. Also acts as an inducer of tumor cytostasis and differentiation. Increases fetal hemoglobin production in vitro and in vivo.
An antineoplastic agent that demonstrates potent differentiating capacity in multiple hematopoietic and solid tumor cell lines. Acts as an inducer of tumor cytostasis and differentiation as well as of peroxisomal proliferation. A more effective inhibitor of histone deacetylase and inducer of histone acetylation than its structural analogs including 2- and 3-phenylbutyrate. Acts as a transcriptional regulator and improve the targeting of δF508-CFTR (cystic fibrosis transmembrane regulator) for ubiquitination and degradation by reducing the expression of HSC70 in epithelial cells. Also, reported to increase fetal hemoglobin production in vitro and in vivo.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Anti-neoplastic agent
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tso, S.C., et al. 2013. Proc. Natl. Acad. Sci. USA110, 9728.
Ozcan, U., et al. 2006. Science313, 1137.
Carducci, M.A., et al. 2001. Clin. Cancer Res.7, 3047.
Rubenstein, R.C. and Zeitlin, P.L. 2000. Am.J. Physiol. Cell. Physiol.278, C259.
Lea, M.A., et al. 1999. Anticancer Res.19, 1971.
Miller, A.C., et al. 1997. Int. J. Radiat. Biol. 72, 211.
Rubenstein, R.C., et al. 1997. J. Clin. Invest. 100, 2457.
Pineau, T., et al. 1996. Biochem. Pharmacol. 52, 659.
Collins, A.F., et al. 1995. Blood 85, 43.
Lea, M.A., et al. 1995. Anticancer Res.15, 879.
Figg, W.D., et al. 1994. Anticancer Drugs 5, 336.
Liu, L., et al. 1994. J. Invest. Dermatol. 103, 335.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uma Maheswari Deshetty et al.
Viruses, 16(6) (2024-06-27)
Antiretroviral treatments have notably extended the lives of individuals with HIV and reduced the occurrence of comorbidities, including ocular manifestations. The involvement of endoplasmic reticulum (ER) stress in HIV-1 pathogenesis raises questions about its correlation with cellular senescence or its
Poonam Aggarwal et al.
Cell reports, 39(12), 110958-110958 (2022-06-23)
We previously showed that the Arf1-mediated lipolysis pathway sustains stem cells and cancer stem cells (CSCs); its ablation resulted in necrosis of stem cells and CSCs, which further triggers a systemic antitumor immune response. Here we show that knocking down
Mariana I Tsap et al.
eLife, 13 (2024-04-25)
Mutations in Drosophila Swiss cheese (SWS) gene or its vertebrate orthologue neuropathy target esterase (NTE) lead to progressive neuronal degeneration in flies and humans. Despite its enzymatic function as a phospholipase is well established, the molecular mechanism responsible for maintaining

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service