Merck
  • Neuroplastin and Basigin Are Essential Auxiliary Subunits of Plasma Membrane Ca2+-ATPases and Key Regulators of Ca2+ Clearance.
Neuroplastin and Basigin Are Essential Auxiliary Subunits of Plasma Membrane Ca2+-ATPases and Key Regulators of Ca2+ Clearance.
Neuron (2017-10-24)
Nadine Schmidt, Astrid Kollewe, Cristina E Constantin, Sebastian Henrich, Andreas Ritzau-Jost, Wolfgang Bildl, Anja Saalbach, Stefan Hallermann, Akos Kulik, Bernd Fakler, Uwe Schulte
ABSTRACT
Plasma membrane Ca2+-ATPases (PMCAs), a family of P-type ATPases, extrude Ca2+ ions from the cytosol to the extracellular space and are considered to be key regulators of Ca2+ signaling. Here we show by functional proteomics that native PMCAs are heteromeric complexes that are assembled from two pore-forming PMCA1-4 subunits and two of the single-span membrane proteins, either neuroplastin or basigin. Contribution of the two Ig domain-containing proteins varies among different types of cells and along postnatal development. Complex formation of neuroplastin or basigin with PMCAs1-4 occurs in the endoplasmic reticulum and is obligatory for stability of the PMCA proteins and for delivery of PMCA complexes to the surface membrane. Knockout and (over)-expression of both neuroplastin and basigin profoundly affect the time course of PMCA-mediated Ca2+ transport, as well as submembraneous Ca2+ concentrations under steady-state conditions. Together, these results establish neuroplastin and basigin as obligatory auxiliary subunits of native PMCAs and key regulators of intracellular Ca2+ concentration.
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Anti-ATP2B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ATP2B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)