Merck
  • Effect of Momordica charantia protein on proliferation, apoptosis and the AKT signal transduction pathway in the human endometrial carcinoma Ishikawa H cell line in vitro.
Effect of Momordica charantia protein on proliferation, apoptosis and the AKT signal transduction pathway in the human endometrial carcinoma Ishikawa H cell line in vitro.
Oncology letters (2017-05-20)
Hang-Zhi Gu, Rong-Rong Lin, Han-Chu Wang, Xue-Jie Zhu, Yan Hu, Fei-Yun Zheng
ABSTRACT
Endometrial carcinoma (EC) is one of the most common female malignancies, and there is an urgent requirement to explore new therapeutic strategies. In the present study, Ishikawa H cells were treated with Momordica charantia protein (MCP30). The cell morphology, growth inhibition rate, cell cycle distribution, and expression of phosphate and tensin homolog, P-AKT and AKT were measured. DNA fragmentation analysis and Annexin V-fluorescein isothiocyanate/propidium iodide double staining assay were used to analyze cell apoptosis. MCP30 decreased the viability of Ishikawa H cells in a dose- and time-dependent manner. The early apoptotic rates of Ishikawa H cells treated with MCP30 at 666.67 pM reached to 16.07±0.15%, following 72 h of treatment. DNA ladder was observed in cells treated with 333.33 and 666.67 pM MCP30 following 72 h of treatment. MCP30 blocks Ishikawa H cells from progressing between the S-phase and the G2/M-phase in a time- and concentration-dependent manner. Western blotting revealed that MCP30 treatment decreased the levels of P-AKT in a dose-dependent manner. It was revealed that MCP30 decreases cell proliferation, and induces apoptosis and S-phase cell cycle arrest through the AKT signaling pathway in Ishikawa H cells.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution