Merck
  • AMPK activation by peri-sciatic nerve administration of ozone attenuates CCI-induced neuropathic pain in rats.
AMPK activation by peri-sciatic nerve administration of ozone attenuates CCI-induced neuropathic pain in rats.
Journal of molecular cell biology (2016-10-17)
Lijuan Lu, Cailong Pan, Lu Chen, Liang Hu, Chaoyu Wang, Yuan Han, Yanjing Yang, Zhixiang Cheng, Wen-Tao Liu
ABSTRACT
Neuropathic pain is a debilitating clinical condition with few efficacious treatments, warranting development of novel therapeutics. Ozone is widely used as an alternative therapy for many different pain conditions, with exact mechanisms still elusive. In this study, we found that a single peri-sciatic nerve injection of ozone decreased mechanical allodynia and thermal hyperalgesia, and normalized the phosphorylation of protein kinase C γ, N-methyl-D-aspartate receptor, and extracellular signal-regulated kinase in a chronic constriction injury (CCI) model in rat sciatic nerve. Meanwhile, ozone significantly suppressed CCI-induced activation of spinal microglia. More importantly, the anti-nociceptive effect of ozone depended on the activation of 5'-adenosine monophosphate (AMP)-activated protein kinase (AMPK), which was proved by the fact that the phosphorylated AMPK level increased during the ozone therapy and AMPK antagonist abolished the effect of ozone in vivo and in vitro. In addition, direct injection of AMPK agonist could replicate the anti-nociceptive effect of ozone in CCI rats. In conclusion, our observations indicate that peri-sciatic nerve injection of ozone activates AMPK to attenuate CCI-induced neuropathic pain.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKAA2
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder