Skip to Content
Merck
All Photos(1)

Key Documents

EHU042081

Sigma-Aldrich

MISSION® esiRNA

targeting human PRKAA2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAGATTCTGTCTGCTGTGGATTACTGTCATAGGCATATGGTTGTTCATCGAGACCTGAAACCAGAGAATGTCCTGTTGGATGCACACATGAATGCCAAGATAGCCGATTTCGGATTATCTAATATGATGTCAGATGGTGAATTTCTGAGAACTAGTTGCGGATCTCCAAATTATGCAGCACCTGAAGTCATCTCAGGCAGATTGTATGCAGGTCCTGAAGTTGATATCTGGAGCTGTGGTGTTATCTTGTATGCTCTTCTTTGTGGCACCCTCCCATTTGATGATGAGCATGTACCTACGTTATTTAAGAAGATCCGAGGGGGTGTCTTTTATATCCCAGAATATCTCAATCGTTCTGTCGCCACTCTCCTGATGCATATGCTGCAGGTTGACCCACTGAAACGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Lin et al.
Frontiers in pharmacology, 10, 1617-1617 (2020-02-13)
The increase of blood pressure accelerates endothelial progenitor cells (EPCs) senescence, hence a significant reduction in the number of EPCs is common in patients with hypertension. Autophagy is a defense and stress regulation mechanism to assist cell homeostasis and organelle
Wei-Wen Su et al.
International journal of medical sciences, 17(5), 678-684 (2020-03-27)
Background: Adenine exhibits potential anticancer activity against several types of malignancies. However, whether adenine has anticancer effects on hepatocellular carcinoma (HCC) cells is incompletely explored. Methods: Human HCC cell lines HepG2 and SK-Hep-1 (p53-wild type) and Hep3B (p53-deficient) were used
Meera Saxena et al.
Journal of cell science, 131(14) (2018-06-29)
The developmental programme of epithelial-mesenchymal transition (EMT), involving loss of epithelial and acquisition of mesenchymal properties, plays an important role in the invasion-metastasis cascade of cancer cells. In the present study, we show that activation of AMP-activated protein kinase (AMPK)
Hua Yu et al.
International journal of oncology, 50(1), 161-172 (2016-12-07)
Caulerpin, a secondary metabolite from the marine invasive green algae Caulerpa cylindracea is known to induce mitochondrial dysfunctions. In this study, the anticancer property of caulerpin was assessed in a panel of colorectal cancer cell lines. We demonstrated that caulerpin
Yingshuai Zhao et al.
Cardiology, 143(1), 1-10 (2019-07-16)
The aberrant proliferation and migration of vascular smooth muscle cells (VSMCs) in the vascular wall are crucial pathological events involved in cardiovascular impairments including hypertension, heart failure, and atherosclerosis. At the molecular level, the mammalian target of rapamycin (mTOR)-ribosomal protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service