Merck
  • Ivermectin Pharmacokinetics, Metabolism, and Tissue/Egg Residue Profiles in Laying Hens.
Ivermectin Pharmacokinetics, Metabolism, and Tissue/Egg Residue Profiles in Laying Hens.
Journal of agricultural and food chemistry (2015-11-11)
Laura Moreno, Paula Dominguez, Cristina Farias, Lucila Canton, Guillermo Virkel, Laura Maté, Laura Ceballos, Carlos Lanusse, Luis Alvarez
ABSTRACT
The goals were to determine the ivermectin (IVM) plasma pharmacokinetics, tissue and egg residue profiles, and in vitro metabolism in laying hens. Experiments conducted were (1) 8 hens were intravenously treated with IVM and blood samples taken; (2) 88 hens were treated with IVM administered daily in water (5 days) (40 were kept and their daily eggs collected; 48 were sacrificed in groups (n = 8) at different times and tissue samples taken and analyzed); (3) IVM biotransformation was studied in liver microsomes. Pharmacokinetic parameters were AUC = 85.1 ng·day/mL, Vdss = 4.43 L/kg, and T1/2el = 1.73 days. Low IVM tissue residues were quantified with the highest measured in liver and skin+fat. IVM residues were not found in egg white, but significant amounts were quantified in yolk. Residues measured in eggs were greater than some MRL values, suggesting that a withdrawal period would be necessary for eggs after IVM use in laying hens.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%