Merck
  • A plasma protein corona enhances the biocompatibility of Au@Fe3O4 Janus particles.
A plasma protein corona enhances the biocompatibility of Au@Fe3O4 Janus particles.
Biomaterials (2015-08-16)
Lisa Landgraf, Carolin Christner, Wiebke Storck, Isabel Schick, Ines Krumbein, Heidi Dähring, Katja Haedicke, Karl Heinz-Herrmann, Ulf Teichgräber, Jürgen R Reichenbach, Wolfgang Tremel, Stefan Tenzer, Ingrid Hilger
ABSTRACT
Au@Fe3O4 Janus particles (JPs) are heteroparticles with discrete domains defined by different materials. Their tunable composition and morphology confer multimodal and versatile capabilities for use as contrast agents and drug carriers in future medicine. Au@Fe3O4 JPs have colloidal properties and surface characteristics leading to interactions with proteins in biological fluids. The resulting protein adsorption layer ("protein corona") critically affects their interaction with living matter. Although Au@Fe3O4 JPs displayed good biocompatibility in a standardized in vitro situation, an in-depth characterization of the protein corona is of prime importance to unravel underlying mechanisms affecting their pathophysiology and biodistribution in vitro and in vivo. Here, we comparatively analyzed the human plasma corona of Au-thiol@Fe3O4-SiO2-PEG JPs (NH2-functionalized and non-functionalized) and spherical magnetite (Fe3O4-SiO2-PEG) particles and investigated its effects on colloidal stability, biocompatibility and cellular uptake. Label-free quantitative proteomic analyses revealed that complex coronas including almost 180 different proteins were formed within only one minute. Remarkably, in contrast to spherical magnetite particles with surface NH2 groups, the Janus structure prevented aggregation and the adhesion of opsonins. This resulted in an enhanced biocompatibility of corona sheathed JPs compared to spherical magnetite particles and corona-free JPs.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
SAFC
Sodium chloride solution, 5 M