Merck
  • Semaphorin 3D autocrine signaling mediates the metastatic role of annexin A2 in pancreatic cancer.
Semaphorin 3D autocrine signaling mediates the metastatic role of annexin A2 in pancreatic cancer.
Science signaling (2015-08-06)
Kelly Foley, Agnieszka A Rucki, Qian Xiao, Donger Zhou, Ashley Leubner, Guanglan Mo, Jennifer Kleponis, Annie A Wu, Rajni Sharma, Qingguang Jiang, Robert A Anders, Christine A Iacobuzio-Donahue, Katherine A Hajjar, Anirban Maitra, Elizabeth M Jaffee, Lei Zheng
ABSTRACT
Most patients with pancreatic ductal adenocarcinoma (PDA) present with metastatic disease at the time of diagnosis or will recur with metastases after surgical treatment. Semaphorin-plexin signaling mediates the migration of neuronal axons during development and of blood vessels during angiogenesis. The expression of the gene encoding semaphorin 3D (Sema3D) is increased in PDA tumors, and the presence of antibodies against the pleiotropic protein annexin A2 (AnxA2) in the sera of some patients after surgical resection of PDA is associated with longer recurrence-free survival. By knocking out AnxA2 in a transgenic mouse model of PDA (KPC) that recapitulates the progression of human PDA from premalignancy to metastatic disease, we found that AnxA2 promoted metastases in vivo. The expression of AnxA2 promoted the secretion of Sema3D from PDA cells, which coimmunoprecipitated with the co-receptor plexin D1 (PlxnD1) on PDA cells. Mouse PDA cells in which SEMA3D was knocked down or ANXA2-null PDA cells exhibited decreased invasive and metastatic potential in culture and in mice. However, restoring Sema3D in AnxA2-null cells did not entirely rescue metastatic behavior in culture and in vivo, suggesting that AnxA2 mediates additional prometastatic mechanisms. Patients with primary PDA tumors that have abundant Sema3D have widely metastatic disease and decreased survival compared to patients with tumors that have relatively low Sema3D abundance. Thus, AnxA2 and Sema3D may be new therapeutic targets and prognostic markers of metastatic PDA.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting human PLXND1
Sigma-Aldrich
L-Glutamine