Merck
  • Dihydrotanshinone I induced apoptosis and autophagy through caspase dependent pathway in colon cancer.
Dihydrotanshinone I induced apoptosis and autophagy through caspase dependent pathway in colon cancer.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2015-11-10)
Lin Wang, Tao Hu, Jing Shen, Lin Zhang, Ruby Lok-Yi Chan, Lan Lu, Mingxing Li, Chi Hin Cho, William Ka Kei Wu
ABSTRACT
Dihydrotanshinone I (DHTS) was previously reported to exhibit the most potent anti-cancer activity among several tanshinones in colon cancer cells. Its cytotoxic action was reactive oxygen species (ROS) dependent but p53 independent. To further study the anti-cancer activity of DHTS and its molecular mechanisms of action in colon cancer both in vitro and in vivo. Caspase activity was detected by fluorescence assay. Apoptosis was detected by flow cytometry and TUNEL assay. Protein levels were analyzed by western blotting. Knockdown of target gene was achieved by siRNA transfection. Formation of LC3B puncta and activation of caspase-3 were detected by confocal fluorescence microscope. In vivo anti-colon cancer activity of DHTS was observed in xenograft tumors in NOD/SCID mice. Anti-colon cancer activity of DHTS by inducing apoptosis and autophagy was observed both in vitro and in vivo. Mitochondria mediated caspase dependent pathway was essential in DHTS-induced cytotoxicity. The apoptosis induced by DHTS was suppressed by knockdown of apoptosis inducing factor (AIF), inhibition of caspase-3/9 but was increased after knockdown of caspase-2. Meantime, knockdown of caspase-2, pretreatment with Z-VAD-fmk or NAC (N-Acety-L-Cysteine) efficiently inhibited the autophagy induced by DHTS. A crosstalk between cytochrome c and AIF was also reported. DHTS-induced caspase and ROS dependent apoptosis and autophagy were mediated by mitochondria in colon cancer. DHTS could be a promising leading compound for the development of anti-tumor agent or be developed as an adjuvant drug for colon cancer therapy.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human PSMD4
Sigma-Aldrich
MISSION® esiRNA, targeting human AF165138.7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Psmd4
Sigma-Aldrich
7-Amino-4-methylcoumarin, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
7-Amino-4-methylcoumarin, Chromophore for substrates
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, ≥97% (HPLC), powder
Sigma-Aldrich
Z-Ile-Glu(O-ME)-Thr-Asp(O-Me) fluoromethyl ketone, ≥90% (TLC), powder