Merck
Swine Dialyzable Spleen Extract as Antiviral Prophylaxis.
Journal of medicinal food (2015-04-14)
Giovanna Merchand-Reyes, Lenin Pavón, Gilberto Pérez-Sánchez, Said Vázquez-Leyva, Nohemí Salinas-Jazmín, Marco Velasco-Velázquez, Emilio Medina-Rivero, Sonia Mayra Pérez-Tapia
ABSTRACT
Worldwide, the most highly consumed meat is of porcine origin. The production and distribution of swine meat are affected by diverse health matters, such as influenza and diarrhea, which cause head losses and require the use of antibiotics and other drugs in hog farms. To stimulate newborn piglet immune responses and increase resistance to infections, we developed a spray-drying technique to produce dried swine dialyzable spleen extract (sDSE), an immunomodulator. Based on the size-exclusion ultra performance liquid chromatography quantitative analysis, it was possible to recover up to 58% of the product after the drying process. The biological activity of orally administered dried sDSE increased mouse survival and induced cytokine production in a herpes infection model.
Product Description
Supelco
Potassium Sorbate, analytical standard
Sigma-Aldrich
Potassium sorbate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
trans,trans-2,4-Hexadienoic acid potassium salt, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%