Merck
Automated, high-throughput serum glycoprofiling platform.
Integrative biology : quantitative biosciences from nano to macro (2015-07-21)
H Stöckmann, R O'Flaherty, B Adamczyk, R Saldova, P M Rudd
ABSTRACT
Complex carbohydrates are rapidly becoming excellent biomarker candidates because of their high sensitivity to pathological changes. However, the discovery of clinical glycobiomarkers has been slow, due to the scarcity of high-throughput glycoanalytical workflows that allow rapid glycoprofiling of large clinical sample sets. To generate high-quality quantitative glycomics data in a high-throughput fashion, we have developed a robotized platform for rapid serum-based N-glycan sample preparation. The sample preparation workflow features a fully automated, rapid glycoprotein denaturation followed by sequential enzymatic glycan release, glycan purification on solid-supported hydrazide and fluorescent labelling. This allows accurate glycan quantitation by ultra-high performance liquid chromatography (UPLC). The sample preparation workflow was automated using an eight-channel Hamilton Robotics liquid handling workstation, allowing the preparation of almost 100 samples in 14 hours with excellent reproducibility and thus should greatly facilitate serum-based glyco-biomarker discovery.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size