Merck
  • Selective distributions of functionalized single-walled carbon nanotubes in a polymeric reverse hexagonal phase.
Selective distributions of functionalized single-walled carbon nanotubes in a polymeric reverse hexagonal phase.
Soft matter (2015-06-11)
Jae-Min Ha, Hyung-Sik Jang, Sung-Hwan Lim, Sung-Min Choi
ABSTRACT
We have investigated the distributions of individually isolated and hydrophilically functionalized single-walled carbon nanotubes (p-SWNTs) in the Pluronic L121-water system at the reverse hexagonal phase using small-angle X-ray scattering (SAXS) and contrast-matched small-angle neutron scattering (SANS) measurements. As the p-SWNT-L121-water system is transitioned from the lamellar phase to the reverse hexagonal phase with temperature, p-SWNTs which were selectively distributed in the polar layers of the lamellar structure become selectively distributed in the cylindrical polar cores of the reverse hexagonal structure, forming a hexagonal array of p-SWNTs. This was clearly confirmed by the contrast-matched SANS measurements. The selective distribution of p-SWNTs in the reverse hexagonal phase is driven by the selective affinity of p-SWNTs to the polar domains of the block copolymer system. The method demonstrated in this study provides a new route for fabricating ordered SWNT superstructures and may be applicable for inorganic 1D nanoparticles such as semiconducting, metallic and magnetic nanorods which are of great interest.
Sigma-Aldrich
4-Vinylbenzoic acid, 97%
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
SAFC
Hexadecyltrimethylammonium bromide, USP/NF
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D