Merck
What drives MRI-measured cortical atrophy in multiple sclerosis?
Multiple sclerosis (Houndmills, Basingstoke, England) (2015-01-15)
V Popescu, R Klaver, P Voorn, Y Galis-de Graaf, D L Knol, J W R Twisk, A Versteeg, G J Schenk, P Van der Valk, F Barkhof, H E De Vries, H Vrenken, J J G Geurts
ABSTRACT
Cortical atrophy, assessed with magnetic resonance imaging (MRI), is an important outcome measure in multiple sclerosis (MS) studies. However, the underlying histopathology of cortical volume measures is unknown. We investigated the histopathological substrate of MRI-measured cortical volume in MS using combined post-mortem imaging and histopathology. MS brain donors underwent post-mortem whole-brain in-situ MRI imaging. After MRI, tissue blocks were systematically sampled from the superior and inferior frontal gyrus, anterior cingulate gyrus, inferior parietal lobule, and superior temporal gyrus. Histopathological markers included neuronal, axonal, synapse, astrocyte, dendrite, myelin, and oligodendrocyte densities. Matched cortical volumes from the aforementioned anatomical regions were measured on the MRI, and used as outcomes in a nested prediction model. Forty-five tissue blocks were sampled from 11 MS brain donors. Mean age at death was 68±12 years, post-mortem interval 4±1 hours, and disease duration 35±15 years. MRI-measured regional cortical volumes varied depending on anatomical region. Neuronal density, neuronal size, and axonal density were significant predictors of GM volume. In patients with long-standing disease, neuronal and axonal pathology are the predominant pathological substrates of MRI-measured cortical volume in chronic MS.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
o-Xylene, anhydrous, 97%