Merck
  • Physiologically generated presenilin 1 lacking exon 8 fails to rescue brain PS1-/- phenotype and forms complexes with wildtype PS1 and nicastrin.
Physiologically generated presenilin 1 lacking exon 8 fails to rescue brain PS1-/- phenotype and forms complexes with wildtype PS1 and nicastrin.
Scientific reports (2015-11-27)
Hannah Brautigam, Cesar L Moreno, John W Steele, Alexey Bogush, Dara L Dickstein, John B J Kwok, Peter R Schofield, Gopal Thinakaran, Paul M Mathews, Patrick R Hof, Sam Gandy, Michelle E Ehrlich
ABSTRACT
The presenilin 1 (PSEN1) L271V mutation causes early-onset familial Alzheimer's disease by disrupting the alternative splicing of the PSEN1 gene, producing some transcripts harboring the L271V point mutation and other transcripts lacking exon 8 (PS1(∆exon8)). We previously reported that PS1 L271V increased amyloid beta (Aβ) 42/40 ratios, while PS1(∆exon8) reduced Aβ42/40 ratios, indicating that the former and not the exon 8 deletion transcript is amyloidogenic. Also, PS1(∆exon8) did not rescue Aβ generation in PS1/2 double knockout cells indicating its identity as a severe loss-of-function splice form. PS1(∆exon8) is generated physiologically raising the possibility that we had identified the first physiological inactive PS1 isoform. We studied PS1(∆exon8) in vivo by crossing PS1(∆exon8) transgenics with either PS1-null or Dutch APP(E693Q) mice. As a control, we crossed APP(E693Q) with mice expressing a deletion in an adjacent exon (PS1(∆exon9)). PS1(∆exon8) did not rescue embryonic lethality or Notch-deficient phenotypes of PS1-null mice displaying severe loss of function in vivo. We also demonstrate that this splice form can interact with wildtype PS1 using cultured cells and co-immunoprecipitation (co-IP)/bimolecular fluorescence complementation. Further co-IP demonstrates that PS1(∆exon8) interacts with nicastrin, participating in the γ-secretase complex formation. These data support that catalytically inactive PS1(∆exon8) is generated physiologically and participates in protein-protein interactions.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
MISSION® esiRNA, targeting human PSEN1
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
SAFC
HEPES
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
SAFC
BIS-TRIS
SAFC
HEPES