Merck
  • Development and evaluation of electromembrane extraction across a hollow polymer inclusion membrane.
Development and evaluation of electromembrane extraction across a hollow polymer inclusion membrane.
Journal of chromatography. A (2015-07-05)
Nor Akma Mamat, Hong Heng See
ABSTRACT
In this work, a new variation of the electromembrane extraction (EME) approach employing a hollow polymer inclusion membrane (HPIM) was developed. In this method, a HPIM was prepared by casting a solution of the desired proportions of cellulose acetate (CTA), tris(2-ethylhexyl)phosphate (TEHP) and di-(2-ethylhexyl)phosphoric acid (D2EHPA) in dichloromethane on glass capillary tubing. Three basic drugs namely amphetamine, methamphetamine, and 3,4-methylenedioxy-N-methylamphetamine (MDMA) were selected as model analytes to evaluate the extraction performance of this new approach. The drugs were extracted from human plasma samples, through a 20μm thickness HPIM, to an aqueous acceptor solution inside the lumen of the hollow membrane. Parameters affecting the extraction efficiency were investigated in detail. Under the optimized conditions, enrichment factors in the range of 97-103-fold were obtained from 3mL of sample solution with a 10min extraction time and an applied voltage of 300V across the HPIM. The detection limits of the method for the three drugs were in the range of 1.0-2.5ng/mL (at a signal/noise ratio of three), with relative standard deviations of between 6.4% and 7.9%. When the method was applied to spiked plasma samples, the relative recoveries ranged from 99.2% to 100.8%. Enrichment factors of 103, 99 and 97 were obtained for amphetamine, methamphetamine, and MDMA, respectively. A comparison was also made between the newly developed approach and EME using supported liquid membranes (SLM) as well as standard sample preparation methods (liquid-liquid extraction) used by the Toxicology Unit, Department of Chemistry, Malaysia.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
1-Chlorobutane, anhydrous, 99.5%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%