Merck
  • Diclofenac-Choline Antioxidant Activity Investigated by means of Luminol Amplified Chemiluminescence of Human Neutrophil Bursts and Electron Paramagnetic Resonance Spectroscopy.
Diclofenac-Choline Antioxidant Activity Investigated by means of Luminol Amplified Chemiluminescence of Human Neutrophil Bursts and Electron Paramagnetic Resonance Spectroscopy.
Drug research (2014-06-12)
P C Braga, N Lattuada, V Greco, V Sibilia, M Falchi, T Bianchi, M Dal Sasso
ABSTRACT
A new diclofenac salt called diclofenac-choline (DC) has recently been proposed for the symptomatic treatment of oropharyngeal inflammatory processes and pain because its greater water solubility allows the use of high concentrations, which are useful when the contact time between the drug and the oropharyngeal mucosa is brief, as in the case of mouthwashes or spray formulations. The antioxidant activity of DC has not yet been investigated, and so the aim was to use luminol-amplified-chemiluminescence (LACL) to verify whether various concentrations of DC (1.48, 0.74 and 0.37 mg/mL for incubation times of 2, 4 and 8 min) interfere with oxygen and nitrogen radicals during the course of human neutrophils respiratory bursts; electron paramagnetic resonance (EPR) spectroscopy was used to investigate its direct antiradical (scavenger) activity. The EPR findings showed that DC has concentration-dependent scavenging activity against the ABTS, the DPPH, and the hydroxyl radicals, but no activity on superoxide anion, as has been previously reported in the case of other NSAIDs. LACL revealed an inhibitory effect that was statistically significant after only 2 min of incubation, and similar after 4 and 8 min. The effects on the peroxynitrite radical paralleled those observed in the previous test. High concentrations and short incubation times showed that there is no interference on PMN viability, and so the inhibitory findings must be attributed to the effect of the drug. The anti-inflammatory effects of DC cannot be attributed solely to the inhibition of prostaglandin synthesis, but its effects on free radicals and neutrophil bursts suggest that they may contribute to its final therapeutic effect.
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
SAFC
L-Arginine
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma