Merck
  • Ovalbumin labeling with p-hydroxymercurybenzoate: The effect of different denaturing agents and the kinetics of reaction.
Ovalbumin labeling with p-hydroxymercurybenzoate: The effect of different denaturing agents and the kinetics of reaction.
Analytical biochemistry (2015-05-07)
Beatrice Campanella, Massimo Onor, Lorenzo Biancalana, Alessandro D'Ulivo, Emilia Bramanti
ABSTRACT
The aim of our study was to investigate how denaturing agents commonly used in protein analysis influence the labeling between a reactive molecule and proteins. For this reason, we investigated the labeling of ovalbumin (OVA) as a globular model protein with p-hydroxymercurybenzoate (pHMB) in its native state (phosphate buffer solution) and in different denaturing conditions (8 molL(-1) urea, 3 molL(-1) guanidinium thiocyanate, 6 molL(-1) guanidinium chloride, 0.2% sodium dodecyl sulfate, and 20% methanol). In addition to chemical denaturation, thermal denaturation was also tested. The protein was pre-column simultaneously denatured and derivatized, and the pHMB-labeled denatured OVA complexes were analyzed by size exclusion chromatography (SEC) coupled online with chemical vapor generation-atomic fluorescence spectrometry (CVG-AFS). The number of -SH groups titrated greatly depends on the protein structure in solution. Indeed, we found that, depending on the adopted denaturing conditions, OVA gave different aggregate species that influence the complexation process. The results were compared with those obtained by a common alternative procedure for the titration of -SH groups that employs monobromobimane (mBBr) as tagging molecule and molecular fluorescence spectroscopy as detection technique. We also investigated the labeling kinetics for denatured OVA and pHMB, finding that the 4 thiolic groups of OVA have a very different reactivity toward mercury labeling, in agreement with previous studies.
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%