Merck
  • Heat Stress Induces Extended Plateau of Hsp70 Accumulation--A Possible Cytoprotection Mechanism in Hepatic Cells.
Heat Stress Induces Extended Plateau of Hsp70 Accumulation--A Possible Cytoprotection Mechanism in Hepatic Cells.
Journal of cellular biochemistry (2015-04-11)
Biljana Miova, Suzana Dinevska-Kjovkarovska, Juan V Esplugues, Nadezda Apostolova
ABSTRACT
The relevance of heat preconditioning resides in its ability to protect cells from different kinds of injury by induction of heat shock proteins, a process in which the intensity of heat stress (HS) and duration of subsequent recovery are vital. This study evaluates the effects of moderate HS (45 min/43°C) and the time-dependent changes during recovery period of HSP70, Bcl-2 and p53 gene and protein expression in HepG2 cells. We also evaluated the effects of 0.4 mM aspirin (ASA) as a potential pharmacological co-inducer of HSP, both alone and in a combination with HS (ASA + HS). HS alone and ASA + HS caused a major up-regulation of HSP70 mRNA in the first 2 h, while HSP70 protein increased gradually and was especially abundant from 2 h to 24 h. Regarding Bcl-2, all treatments rendered similar results: gene expression was down-regulated in the first 2 h, after which there was protein elevation (12-48 h after HS). mRNA expression of p53 in HS- and (ASA + HS)-cells was down-regulated in the first 12 h. The immediate decrease of p53 protein after HS was followed by a biphasic increase. In conclusion, 0.4 mM ASA + HS does not act as a co-inducer of HSP70 in HepG2 cells, but promotes Bcl-2 protein expression during prolonged treatment. Our suggestion is that hepatic cells are most vulnerable in the first 2-6 h, but may have a high capacity for combating stress 12-24 h after HS. Finally, short-term exposure HS might be a "physiological conditioner" for liver cells to accumulate HSP and Bcl-2 proteins and thus obtain cytoprotection against an additional stress.
Sigma-Aldrich
Anti-p53, antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Anti-Hsp70 Mouse mAb (C92F3A-5), liquid, clone C92F3A-5, Calbiochem®
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%