Merck
  • Phenolic Profiling of Olives and Olive Oil Process-Derived Matrices Using UPLC-DAD-ESI-QTOF-HRMS Analysis.
Phenolic Profiling of Olives and Olive Oil Process-Derived Matrices Using UPLC-DAD-ESI-QTOF-HRMS Analysis.
Journal of agricultural and food chemistry (2015-03-19)
Tina Jerman Klen, Alenka Golc Wondra, Urška Vrhovšek, Branka Mozetič Vodopivec
ABSTRACT
All of the matrices entailed in olive oil processing were screened for the presence of known and new phenol constituents in a single study, combining an ultra high pressure liquid chromatography system with diode array and electrospray ionization quadrupole time-of-flight high resolution mass spectrometry (ESI-QTOF-HRMS) detection. Their trail was followed from the fruit (peel/pulp and stone) to the paste and final products, i.e. pomace, wastewater, and oil, providing important insight into the origin, disappearance, and evolution of each during the operational steps. Eighty different phenols, composed of fruit native representatives and their technologically formed and/or released derivatives, were detected in six olive matrices and fully characterized on the basis of HRMS and UV-vis spectroscopic data. In addition to phenols already known in olive matrices, four new molecular formulas were proposed and three new tentative identities assigned to newly discovered phenols, i.e., β-methyl-OH-verbascoside, methoxynüzhenide, and methoxynüzhenide 11-methyl oleoside.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Pinoresinol, ≥95.0% (HPLC)
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
2-(4-Hydroxyphenyl)ethanol, 98%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)