Merck
  • MiR-570 inhibited the cell proliferation and invasion through directly targeting B7-H1 in hepatocellular carcinoma.
MiR-570 inhibited the cell proliferation and invasion through directly targeting B7-H1 in hepatocellular carcinoma.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-06-19)
Wei Guo, Wei Tan, Shan Liu, Xuhui Huang, Juze Lin, Ronghua Liang, Le Su, Qiao Su, Changjun Wang
ABSTRACT
A recent study reported that miR-570 was the most important microRNA in the microRNA gene networks of alcoholic liver disease that has the potential of progressing to hepatocellular carcinoma. However, litter is known regarding the expression and specific function of miR-570 in the progression of hepatocellular carcinoma, especially its molecular mechanisms by which miR-570 exerts its functions and modulates the malignant phenotypes of hepatocellular carcinoma cells. Here, we observed that miR-570 was highly expressed in hepatocellular carcinoma cell lines (Bel-7404, Huh-7, and HepG2), while B7-H1 was lowly expressed, compared to nonmalignant cell line (L-02 and HL-7702). Transfection of miR-570 mimics or knockdown of B-H1 suppressed the expression of B7-H1, which promotes cell apoptosis and inhibits the cell proliferation and invasion. Using a dual-luciferase reporter system, we verified that B7-H1 is a direct target of miR-570. The overexpression of B7-H1 reversed the inhibition of proliferation and invasion by miR-570. In addition, miR-570 suppressed tumorigenicity in vivo. Hence, our observation confirmed that miR-570 works as proliferation and metastatic suppressor in hepatocellular carcinoma cells through directly targeting B7-H1 in hepatocellular carcinoma cell and rationally presents that miR-570 has the potential to be a useful clinical noninvasive diagnostics or predictive marker in human hepatocellular carcinoma.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human CLIC4
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
MISSION® esiRNA, targeting human HIST1H1B
Sigma-Aldrich
MISSION® esiRNA, targeting human RLN1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Clic4
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rln1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Vps45
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting human VPS45