Merck
  • Magnetic graphene as modified quick, easy, cheap, effective, rugged and safe adsorbent for the determination of organochlorine pesticide residues in tobacco.
Magnetic graphene as modified quick, easy, cheap, effective, rugged and safe adsorbent for the determination of organochlorine pesticide residues in tobacco.
Journal of chromatography. A (2015-06-21)
Yan-Bo Luo, Xue Li, Xing-Yi Jiang, Bao-Dong Cai, Feng-Peng Zhu, Hong-Fei Zhang, Zai-Gen Chen, Yong-Qiang Pang, Yu-Qi Feng
ABSTRACT
In this study, magnetic graphene was used as modified quick, easy, cheap, effective, rugged and safe (QuEChERS) adsorbent for the determination of organochlorine pesticide (OCPs) residues in tobacco. To achieve the optimum conditions of modified QuEChERS procedure toward target analytes, several parameters affecting the clean-up efficiency including the amount of the adsorbent and clean-up time were investigated. Under the optimized conditions, a method for the determination of 26 OCPs residues in tobacco was established by coupling the modified QuEChERS procedure to on-line gel permeation chromatography-gas chromatography-tandem mass spectrometry (on-line GPC-GC-MS(2)). The limits of detection of proposed method for 26 OCPs residues ranged from 0.01275 to 3.150ng/g. And good linearities of the proposed method were obtained with coefficients of determination (R(2)) greater than 0.9985 for all target analytes. Good reproducibility of method was obtained as intra- and inter-day precisions, the relative standard deviations were less than 11.1 and 15.0%, respectively. The apparent recoveries were in the range of 64-126% at different concentrations for real samples. Compared with the reported methods for the determination of OCPs residues in tobacco, the proposed method has the advantages of simple to operate, low cost and high clean-up ability. Finally, the method was successfully applied to the analysis of OCPs residues in real samples.
Sigma-Aldrich
Potassium permanganate, meets USP testing specifications
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Citric acid monohydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, E330, 99.5-100.5% (based on anhydrous substance), grit
Sigma-Aldrich
Citric acid monohydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Triphenyl phosphate, ≥99%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Citric acid monohydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Citric acid monohydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)
Sigma-Aldrich
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis