Merck
Wnt pathway in atypical teratoid rhabdoid tumors.
Neuro-oncology (2014-09-24)
Madhavi Chakravadhanula, Chris N Hampton, Parth Chodavadia, Victor Ozols, Li Zhou, Daniel Catchpoole, Jingying Xu, Anat Erdreich-Epstein, Ratan D Bhardwaj
ABSTRACT
Atypical teratoid rhabdoid tumor (ATRT) is an aggressive pediatric brain tumor with limited therapeutic options. The hypothesis for this study was that the Wnt pathway triggered by the Wnt5B ligand plays an important role in ATRT biology. To address this hypothesis, the role of WNT5B and other Wnt pathway genes was analyzed in ATRT tissues and ATRT primary cell lines. Transcriptome-sequencing analyses were performed using nanoString platforms, immunohistochemistry, Western blotting, quantitative reverse transcriptase PCR, immunoprecipitation, short interference RNA studies, cell viability studies, and drug dose response (DDR) assays. Our transcriptome-sequencing results of Wnt pathway genes from ATRT tissues and cell lines indicated that the WNT5B gene is significantly upregulated in ATRT samples compared with nontumor brain samples. These results also indicated a differential expression of both canonical and noncanonical Wnt genes. Imunoprecipitation studies indicated that Wnt5B binds to Frizzled1 and Ryk receptors. Inhibition of WNT5B by short interference RNA decreased the expression of FRIZZLED1 and RYK. Cell viability studies a indicated significant decrease in cell viability by inhibiting Frizzled1 receptor. DDR assays showed promising results with some inhibitors. These promising therapeutic options will be studied further before starting a translational clinical trial. The success of these options will improve care for these patients.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human WNT5B
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
SAFC
BIS-TRIS