Merck
  • Stability-indicating UPLC method for determining related substances and degradants in dronedarone.
Stability-indicating UPLC method for determining related substances and degradants in dronedarone.
Journal of chromatographic science (2013-07-19)
Surya Prakash Rao Pydimarry, Vijay Kumar Cholleti, Ranga Reddy Vangala
ABSTRACT
A simple, sensitive and reproducible method was developed on ultra-performance liquid chromatography coupled with photodiode array detection for the quantitative determination of dronedarone hydrochloride (DRO) in drug substance and pharmaceutical dosage forms. The method is applicable for the quantification of related substances and assays of drug substances. Chromatographic separation was achieved on Acquity UPLC BEH C8 100 mm, 2.1 mm and 1.7 µm columns, using gradient elution within a short run time of 10.0 min. The eluted compounds were monitored at 288 nm, the flow rate was 0.5 mL/min and the column oven temperature was maintained at 40°C. The resolution of DRO and 11 impurities (potentials and by-products) was greater than 2.0 for all pairs of components. The high correlation coefficient value (>0.9995) indicates the clear correlations between the concentrations of investigated compound and their peak areas within the test ranges. The repeatability and intermediate precision, expressed by the relative standard deviation, were less than 2.5%. The accuracy and validity of the method were further ascertained by performing recovery studies via a spike method. The accuracy of the method, expressed as relative error, was satisfactory. No interference was observed from concomitant substances normally added to the tablets. DRO was subjected to the stress conditions of oxidative, acid, base, hydrolytic, thermal and photolytic degradation. DRO was found to degrade significantly in acid and base stress conditions and to remain stable in thermal, photolytic degradation, oxidative and hydrolytic conditions. The degradation products were well resolved from primary peak and its impurities, proving that the method is stability indicating. The developed method was validated as per International Conference on Harmonization guidelines with respect to specificity, limit of detection, limit of quantification, linearity, accuracy, precision, solution stability and robustness. This method is also suitable for the determination of DRO drug substance and pharmaceutical dosage forms.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, anhydrous, free-flowing, Redi-Dri, 97%