Merck
  • Identification of Furosemide Photodegradation Products in Water-Acetonitrile Mixture.
Identification of Furosemide Photodegradation Products in Water-Acetonitrile Mixture.
Chemical & pharmaceutical bulletin (2015-08-04)
Shinji Katsura, Nobuo Yamada, Atsushi Nakashima, Sumihiro Shiraishi, Takayuki Furuishi, Haruhisa Ueda
ABSTRACT
The aim of this study was to identify the chemical structure of the photodegradation products of furosemide in a water-acetonitrile mixture (1 : 1). Furosemide solution was irradiated with a D65 fluorescent lamp and the products were isolated by preparative HPLC. The fractions were evaporated to dryness in vacuo. The purity of the photodegradation products was measured by HPLC. The purity of products 1, 3, and 4 was greater than 90%, whereas that of product 2 was 13%, therefore, photodegradation product 2 was unstable. We identified photodegradation products 1 and 3 as 4-chloro-5-sulfamoylanthranilic acid and 4-hydroxy-N-furfuryl-5-sulfamoylanthranilic acid, respectively, by LC/MS and NMR. Additionally, we assumed that photodegradation product 4 was methyl 2-((furan-2-ylmethyl)amino)-4-hydroxy-3-(methyleneamino)-5-sulfamoylbenzoate by LC/MS and NMR. This showed that furosemide underwent hydrolysis and substitution, and reacted with the acetonitrile under the light of a D65 fluorescent lamp. We were furthermore able to determine the elution times of the photodegradation products of furosemide by applying the Japanese Pharmacopoeia chromatographic method for related substances to the isolated products.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Furosemide
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG